missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
2A Peptide Monoclonal antibody specifically detects 2A Peptide in Human, Nonspecies specific samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | 2A Peptide |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3H4R |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 2A cleavage peptide, 2A Peptide, 2A peptide tag, 2A sequence, 2A-like peptide, antibody for 2A-containing recombinant proteins, FMDV 2A-peptide, MDV 2A-peptide, P2A peptide, P2A tagged proteins, peptide 2A sequences, self-cleaving 2A peptide, T2A peptide, T2A tagged proteins, Thosea asigna virus 2A |
| Host Species | Mouse |
| Immunogen | This Recombinant 2A Peptide Antibody (3H4) was developed against KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?