missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ 293T Whole Cell Lysate (Denatured)

Product Code. 16089645
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 16089645

Brand: Abnova™ L151V1.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Cell Line Lysate

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Tissue Kidney
Description Whole cell lysate (denatured)
Lysis Buffer 1X Sample Buffer (0.2 M Tris-HCl, 2% SDS, 10% glycerol, 0.2% 2-mercaptoethanol, 0.2% Bromophenol blue).
Preparation Method Culture cells in 6-well plate to 80% confluent, remove media and then rinse with cold 1x PBS. Scrape cells off the plate surface and pipette cells into microcentrifuge tube to spin down the cells. Remove PBS and then lysis and resuspend cells with 205 ul 1X sample buffer. Mix well and heat at 90°C for 10 minutes, and then cooled rapidly.
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 100 μL
Storage Buffer In ready-to-use 1X Sample Buffer (0.2 M Tris-HCl, 2% SDS, 10% glycerol, 0.2% 2-mercaptoethanol, 0.2% Bromophenol blue).
Host Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL 293T whole cell lysate (denatured)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P233-Keep container tightly closed.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2-mercaptoethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.