Learn More
IDEXX Water 290 mL Water Sample Vessel
IDEXX Water 290 mL Water Sample Vessel is a versatile vessel that can be used for bottled water samples or larger volumes intended for multiple tests.
Description
Description
  • Shrink-banded
  • Disposable
  • Shrink band acts a tamper-evident seal
  • Protects the entire perimeter from possible contamination
  • High-clarity and nonfluorescing polystyrene helps you see color and fluorescent end points clearly
  • Certified sterilization: Eliminate false-positive results due to contamination
  • Volume lines around entire perimeter
  • Embossed catalog numbers and lot numbers on the bottom of each vessel