missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMS™ Series 1A Model 280 Cooled Incubator, 272 L, Pre-coated Aluminum
Description
- Range of cooled incubators with a white stove enamel steel exterior, foam insulation and a white pre-coated aluminum interior
- Reliable and accurate temperature control
- Standard and programmable models available
- Programmable models offer multiple temperature control in multiple time zones
- Temperature range -10.0°C to +50.0°C, temperature variation with average workload ±0.5°C
- Self-evaporating condensate drip tray
- White plastic coated wire shelves
- Energy efficient and eco-friendly
- P.I.D. heating and cooling to ensure stable internal temperature
- Pre-set safety cutout
- Hermetically sealed refrigeration system
- Door lock
- Controls are easy to read and simple to use
- Full range of optional extras available. including LED lighting, access porthole, inner perspex doors, alarm system, internal socket and chart recorder
- Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 |
| Controller Type | Standard digital |
| Description | Freestanding incubator |
| Capacity (Metric) | 272 L |
| Chamber Material | Pre-coated Aluminum |
| Convection Type | Fan-assisted Circulation |
| Dimensions (D x W) Shelf | 420 mm x 386 mm |
| Lighting | Optional LED |
| Material | Stove Enameled Mild Steel |
| No. of Shelves | 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
