missing translation for 'onlineSavingsMsg'
Learn More

28-Epirapamycin, TRC

Product Code. 30375970
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375970 2.5 mg
30369898 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375970 Supplier Toronto Research Chemicals Supplier No. TRCE5886802.5MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253431-35-5
Molecular Formula C51H79NO13
Molecular Weight (g/mol) 914.17
Synonym 28-epi-Rapamycin, (31S)-Rapamycin
SMILES [H][C@]1(C(O[C@@]([H])(CC([C@H](C)/C=C([C@H](O)[C@H]2OC)\C)=O)[C@@H](C[C@@]3([H])C[C@@H](OC)[C@H](O)CC3)C)=O)N(C(C([C@@]4(O)[C@H](C)CC[C@@]([H])(C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C2=O)O4)=O)=O)CCCC1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C51H79NO13/c1-30-16-12-11-13-17-31(2)42(61-8)28-38-21-19-36(7)51(60,65-38)48(57)49(58)52-23-15-14-18-39(52)50(59)64-43(33(4)26-37-20-22-40(53)44(27-37)62-9)29-41(54)32(3)25-35(6)46(56)47(63-10)45(55)34(5)24-30/h11-13,16-17,25,30,32-34,36-40,42-44,46-47,53,56,60H,14-15,18-24,26-29H2,1-10H3
Formula Weight 913.5551
Chemical Name or Material 28-Epirapamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.