missing translation for 'onlineSavingsMsg'
Learn More
Learn More
26S proteasome subunit 9 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
26S proteasome subunit 9 Polyclonal antibody specifically detects 26S proteasome subunit 9 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Gene Alias | 26S proteasome non-ATPase regulatory subunit 9, p27, proteasome (prosome, macropain) 26S subunit, non-ATPase, 9, Rpn4 |
| Purification Method | Antigen and protein A Affinity-purified |
| Regulatory Status | RUO |
| Research Discipline | Breast Cancer, Cancer, Tumor Suppressors |
| Gene ID (Entrez) | 5715 |
| Target Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?