missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-MSK1(S376)/MSK2(S360) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB1094-SP
This item is not returnable.
View return policy
Description
MSK1/MSK2 Monoclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| MSK1/MSK2 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB | |
| Phosphopeptide containing the human MSK1(S376) site. Accession # O75582 | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 1013D | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL | |
| O75582 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively. | |
| 12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction