missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Phospho-PLC-gamma 1 (Y783) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB74541-SP
This item is not returnable.
View return policy
Description
PLC-gamma 1 Monoclonal specifically detects PLC-gamma 1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PLC-gamma 1 (p tyr783) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLCG1, PLCgamma 1, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PPLCA, triphosphoinositide phosphodiesterase | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 5335 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 1016A | |
| Western Blot 0.05 ug/mL | |
| P19174 | |
| PLCG1 | |
| Phosphopeptide containing the human/mouse/rat PLC-gamma 1 Y783 site Accession # P19174 | |
| 25 μg | |
| Primary | |
| Detects human/mouse/rat PLC-gamma 1 when phosphorylated at Y783 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction