missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Synaptotagmin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4364-SP
This item is not returnable.
View return policy
Description
Synaptotagmin 1 Monoclonal specifically detects Synaptotagmin 1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| Synaptotagmin 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL | |
| DKFZp781D2042, p65, SVP65, synaptotagmin Isynaptotagmin 1, synaptotagmin-1, Synaptotagmin1, SYT, SYT1, SytI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6857 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| ASV48 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SYT1 | |
| Rat brain synaptic plasma membranes | |
| 25 μg | |
| Primary | |
| Detects Synaptotagmin-1 from rat, mouse, human, chicken, Xenopus, and fish. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction