missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PlGF-4 Protein

Product Code. 18738827
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18738827

Brand: R&D Systems™ 8987PL025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human PlGF-4 Protein is derived from HEK293. The Recombinant Human PlGF-4 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49763-4
For Use With (Application) Bioactivity
Gene ID (Entrez) 5228
Molecular Weight (g/mol) Observed M.W.: 35-40kDa, reducing conditions, Theoretical M.W.: 25kDa
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human PlGF-4 protein Leu19-Arg242
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name PlGF-4
Gene Symbol PGF
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R1/Flt-1 Fc Chimera, aa 27-328 is immobilized at 0.5μg/mL, 100μL/well, the concentration of Recombinant Human PlGF-4 that produces 50% of the optimal binding response is approximately 4-24ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Leu19-Arg242
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.