missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat SP-D Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6839-SP
This item is not returnable.
View return policy
Description
SP-D Polyclonal specifically detects SP-D in Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| SP-D | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6441 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| P50404 | |
| SFTPD | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse SP-D Ala20-Phe374 Accession # P50404 | |
| 25 μg | |
| Primary | |
| Detects mouse and rat SP-D in Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human SP-D is observed. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction