missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Semaphorin 4C Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB8497-SP
This item is not returnable.
View return policy
Description
Semaphorin 4C Monoclonal specifically detects Semaphorin 4C in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Semaphorin 4C | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMA4C, SEMACL1, SEMAF, SEMAI, semaphorin-4C | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54910 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 663203 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q64151 | |
| SEMA4C | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4C Ala21-Phe664 Accession # Q64151 | |
| 25 μg | |
| Primary | |
| Detects mouse Semaphorin 4C in direct ELISAs. Detects HEK293 human embryonic kidney cell line transfected with mouse Semaphorin 4C by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction