Learn More
Mouse S100A4 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB4138-SP
Product Code. 18733989
  • £168.00 / 25µg
Description
S100A4 Monoclonal specifically detects S100A4 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S100A4
Monoclonal
LYOPH
Western Blot 2 ug/mL
P07091
S100A4
E. coli-derived recombinant mouse S100A4 Ala2-Lys101 Accession # P07091
25 μg
Primary
Detects mouse S100A4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse S100A6, 8, 9, 10, 13, 16, recombinant human (rh) S100A1, 2, 7, 11, or rhS100P is observed.
Mouse
Purified
Western Blot
438709
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
18A2, 42A, Calvasculin, CAPL, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog)
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
6275
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b