missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pentraxin 3/TSG-14 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2166-SP
This item is not returnable.
View return policy
Description
Pentraxin 3/TSG-14 Polyclonal specifically detects Pentraxin 3/TSG-14 in Mouse samples. It is validated for Western Blot.
Specifications
| Pentraxin 3/TSG-14 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5806 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| P48759 | |
| PTX3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Pentraxin 3/TSG-14 Glu18-Ser381 Accession # P48759 | |
| 25 μg | |
| Primary | |
| Detects mouse Pentraxin 3/TSG-14 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human Pentraxin 3 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction