Learn More
Mouse Protein S/PROS1 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB4976-SP
Product Code. 18792578
  • £184.00 / 25µg
Description
Protein S/PROS1 Monoclonal specifically detects Protein S/PROS1 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein S/PROS1
Monoclonal
LYOPH
Western Blot 2 ug/mL
Q08761
PROS1
Chinese hamster ovary cell line CHO-derived recombinant mouse Protein S/PROS1 Met1-Arg669 Accession # Q08761
25 μg
Primary
Detects mouse Protein S/PROS1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Protein S is observed.
Mouse
Purified
Western Blot
818002
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 3.4.21, PROS, PROS1, protein S (alpha), protein Sa, PS21, PS22, PS23, PS24, PS25, PSA, vitamin K-dependent plasma protein S, vitamin K-dependent protein S
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
5627
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis
For Research Use Only