missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lefty-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7648-SP
This item is not returnable.
View return policy
Description
Lefty-2 Monoclonal specifically detects Lefty-2 in Mouse samples. It is validated for Immunocytochemistry.
Specifications
| Lefty-2 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P57785 | |
| LEFTY2 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Lefty-2 Phe78-Leu368 Accession # P57785 | |
| 25 μg | |
| Primary | |
| Detects mouse Lefty-2 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Lefty-A or recombinant mouse Lefty-1 is observed. | |
| Mouse | |
| Purified |
| Immunocytochemistry | |
| 836101 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EBAF, Endometrial Bleeding-Associated Factor, LEFTA, Left-Right Determination Factor 2, Left-right determination factor A, LEFTYA, Protein Lefty-2, Protein Lefty-A, TGFB4, TGF-beta 4, TGF-beta-4, Transforming Growth Factor Beta-4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 320202 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction