Learn More
Mouse IL-31 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB3028-SP
 View more versions of this product
Product Code. 18772597
  • £167.00 / 25µg
Description
IL-31 Monoclonal specifically detects IL-31 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-31
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q6EAL8
IL31
E. coli-derived recombinant mouse IL-31 Thr24-Cys163 Accession # Q6EAL8
25 μg
Primary
Detects mouse IL-31 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-31.
Mouse
Purified
Western Blot
323407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IL31, IL-31interleukin-31, interleukin 31
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
386653
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b