missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Endoglin/CD105 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB1320-SP
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| NP_031958 | |
| ENG | |
| Mouse myeloma cell line NS0-derived recombinant mouse Endoglin Glu21-Gly581 (predicted) Accession # NP_031958 | |
| 25 μg | |
| Primary | |
| Detects mouse Endoglin in direct ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant human Endoglin. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 209701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2022 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction