missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Chordin-like 2/CHRDL2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2520-SP
This item is not returnable.
View return policy
Description
Chordin-like 2/CHRDL2 Monoclonal specifically detects Chordin-like 2/CHRDL2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Chordin-like 2/CHRDL2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8VEA6.1 | |
| CHRDL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Chordin-like 2/CHRDL2 Gln24-Leu426 Accession # Q8VEA6.1 | |
| 25 μg | |
| Primary | |
| Detects mouse Chordin-like 2/CHRDL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CHRDL2, recombinant mouse (rm) CHRDL1, or rmChordin is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 328905 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BNF-1, Chordin like 2, chordin-like 2, chordin-like protein 2, CHRDL2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25884 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction