missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Aminoacylase/ACY1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2700-SP
This item is not returnable.
View return policy
Description
Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Aminoacylase/ACY1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NP_079647 | |
| ACY1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Aminoacylase/ACY1 Met1-Ser408 Accession # NP_079647 | |
| 25 μg | |
| Primary | |
| Detects human Aminoacylase/ACY1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human ACY1 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 475730 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACY1, ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 95 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction