missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat HO-1/HMOX1/HSP32 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3169-SP
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Polyclonal specifically detects HO-1/HMOX1/HSP32 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HO-1/HMOX1/HSP32 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HMOX1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3162 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P06762 | |
| HMOX1 | |
| E. coli-derived recombinant rat HO-1/HMOX1 Met1-Met289 Accession # P06762 | |
| 25 μg | |
| Primary | |
| Detects mouse and rat HO-1/HMOX1/HSP32 in Western blots. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction