missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREML1/TLT-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2394-SP
This item is not returnable.
View return policy
Description
TREML1/TLT-1 Monoclonal specifically detects TREML1/TLT-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| TREML1/TLT-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q86YW5 | |
| TREML1 | |
| Mouse myeloma cell line NS0-derived recombinant human TREML1/TLT-1 Gln16-Ser160 Accession # Q86YW5 | |
| 25 μg | |
| Primary | |
| Detects human TREML1/TLT-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse TREML1 or recombinant human TREML2 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 268420 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dJ238O23.3, GLTL1825, PRO3438, TLT1, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 340205 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction