missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic Sulfotransferase 1B1/SULT1B1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5959-SP
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1B1/SULT1B1 Monoclonal specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Cytosolic Sulfotransferase 1B1/SULT1B1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O43704 | |
| SULT1B1 | |
| E. coli-derived recombinant human Cytosolic Sulfotransferase 1B1/SULT1B1 Leu2-Ile296 Accession # O43704 | |
| 25 μg | |
| Primary | |
| Detects human Cytosolic Sulfotransferase 1B1/SULT1B1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) SULT1A1, rhSULT1C1, or rhSULT1E1 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 724429 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B1, SULT1B2, Thyroid hormone sulfotransferase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27284 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction