missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STRA6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5904-SP
This item is not returnable.
View return policy
Description
STRA6 Monoclonal specifically detects STRA6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| STRA6 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| AAH25256 | |
| STRA6 | |
| NS0 mouse myeloma cell line transfected with human STRA6 Met1-Pro667 Accession # AAH25256 | |
| 25 μg | |
| Primary | |
| Detects human STRA6 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 496613 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ12541, MCOPS9, stimulated by retinoic acid gene 6 homolog (mouse), stimulated by retinoic acid gene 6 protein homolog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64220 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction