Learn More
Human SLP-76/LCP2 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB5637-SP
Product Code. 18761069
  • £157.00 / 25µg
Description
SLP-76/LCP2 Monoclonal specifically detects SLP-76/LCP2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLP-76/LCP2
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q13094
LCP2
E. coli-derived recombinant human SLP-76 Met1-Pro533 Accession # Q13094
25 μg
Primary
Detects endogenous human SLP-76 in Western blots.
Human
Purified
Western Blot
460004
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
LCP2, lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP76, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3937
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1