missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-5/CD170 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB1072-SP
This item is not returnable.
View return policy
Description
Siglec-5/CD170 Monoclonal specifically detects Siglec-5/CD170 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Siglec-5/CD170 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O15389 | |
| SIGLEC5 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-5/CD170 Lys18-Thr434 Accession # O15389 | |
| 25 μg | |
| Primary | |
| Detects human Siglec-5/CD170 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -7, -9, or -14 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 194117 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD170, CD170 antigen, CD33 antigen-like 2, CD33L2, CD33L2OB binding protein-2, OBBP2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, Siglec5, SIGLEC-5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8778 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction