missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human R-Spondin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3500-SP
This item is not returnable.
View return policy
Description
R-Spondin 3 Polyclonal specifically detects R-Spondin 3 in Human samples. It is validated for Western Blot.
Specifications
| R-Spondin 3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84870 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9BXY4 | |
| RSPO3 | |
| Mouse myeloma cell line NS0-derived recombinant human R-Spondin 3 Ser21-Gly209 Accession # Q9BXY4 | |
| 25 μg | |
| Primary | |
| Detects human R-Spondin 3 in direct ELISAs and Western blots. In direct ELISAs, less than 50% cross-reactivity with recombinant mouse R-Spondin 3 is observed and less than 2% cross-reactivity with recombinant human R-Spondin 1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction