missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin B3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4958-SP
This item is not returnable.
View return policy
Description
Plexin B3 Monoclonal specifically detects Plexin B3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Plexin B3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLEXB3, plexin B3, plexin-B3, PLEXR, PLXN6, PLXNB3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5365 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 905805 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9ULL4 | |
| PLXNB3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Plexin B3 His45-Gln1255 Accession # Q9ULL4 | |
| 25 μg | |
| Primary | |
| Detects human Plexin B3 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction