missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Perilipin-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7664-SP
This item is not returnable.
View return policy
Description
Perilipin-3/TIP47 Monoclonal specifically detects Perilipin-3/TIP47 in Human samples. It is validated for Western Blot.
Specifications
| Perilipin-3/TIP47 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O60664 | |
| PLIN3 | |
| E. coli-derived recombinant human Perilipin-3 Met1-Lys434 Accession # O60664 | |
| 25 μg | |
| Primary | |
| Detects human Perilipin-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Perilipin-2 or rhPerilipin-5 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 779119 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cargo selection protein TIP47, M6PRBP1, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Perilipin3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PLIN3, PP17, PP17MGC11117, tail-interacting protein, 47 kD, TIP47, TIP47mannose-6-phosphate receptor binding protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10226 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction