missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat N-WASP Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3854-SP
This item is not returnable.
View return policy
Description
N-WASP Polyclonal specifically detects N-WASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| N-WASP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NWASP, N-WASPneural WiskNWASP, TRS4, WASL, WiskDKFZp779G0847, WiskMGC48327 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8976 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| O00401 | |
| WASL | |
| E. coli-derived recombinant human N-WASP Leu21-Asn270 Accession # O00401 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat N-WASP in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction