missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MafG Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3924-SP
This item is not returnable.
View return policy
Description
MafG Monoclonal specifically detects MafG in Human samples. It is validated for Western Blot.
Specifications
| MafG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O15525 | |
| MAFG | |
| E. coli-derived recombinant human MafG Met1-Ser162 Accession # O15525 | |
| 25 μg | |
| Primary | |
| Detects human MafG in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) MafF or rhMafK is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 370412 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basic leucine zipper transcription factor MafG, hMAF, MGC13090, MGC20149, transcription factor MafG, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein G, V-maf musculoaponeurotic fibrosarcoma oncogene homolog G, v-maf musculoaponeurotic fibrosarcoma oncogene homolog G (avian) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4097 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction