missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IKK beta Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4535-SP
This item is not returnable.
View return policy
Description
IKK beta Polyclonal specifically detects IKK beta in Human samples. It is validated for Western Blot.
Specifications
| IKK beta | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKB, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3551 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O14920 | |
| IKBKB | |
| E. coli-derived recombinant human IKK beta Gly374-Ser756 Accession # O14920 | |
| 25 μg | |
| Primary | |
| Detects endogenous human IKK beta in Western blots. In Western blots, this antibody does not cross-react with recombinant human IKK alpha , IKK gamma , or IKK epsilon . | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction