missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chorionic Gonadotropin alpha Chain ( alpha HCG) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4169-SP
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Chorionic Gonadotropin alpha Chain (HCG alpha) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P01215 | |
| CGA | |
| Mouse myeloma cell line NS0-derived recombinant human Chorionic Gonadotropin, alpha Chain ( alpha HCG) Ala25-Ser116 Accession # P01215 | |
| 25 μg | |
| Primary | |
| Detects human Chorionic Gonadotropin, alpha Chain ( alpha HCG) in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human FSH beta or recombinant rat FSH beta is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 381012 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha HCG, CGA, CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHa1, HCG, LHA, TSHA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1081 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction