missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLI-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3324-SP
This item is not returnable.
View return policy
Description
GLI-1 Polyclonal specifically detects GLI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP).
Specifications
| GLI-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GLI family zinc finger 1, GLI1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2735 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ChIP Assay, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/10^6 cells | |
| P08151 | |
| GLI1 | |
| E. coli-derived recombinant human GLI-1 Met1-Glu234 Accession # P08151 | |
| 25 μg | |
| Primary | |
| Detects human GLI-1 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human GLI-3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction