missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Growth Hormone R/GHR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1210-SP
This item is not returnable.
View return policy
Description
Growth Hormone R Monoclonal specifically detects Growth Hormone R in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Growth Hormone R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.025-0.1 ug/mL | |
| P10912 | |
| GHR | |
| Mouse myeloma cell line NS0-derived recombinant human Growth Hormone R/GHR Phe19-Tyr264 Accession # P10912 | |
| 25 μg | |
| Primary | |
| Detects human Growth Hormone R/GHR in Western blots and direct ELISAs. In Western blots, no cross-reactivity with recombinant mouse GHR or recombinant rat GHR is observed. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| 193804 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GH receptor, GHBP, GHR, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2690 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction