missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic beta-Glucosidase/GBA3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5969-SP
This item is not returnable.
View return policy
Description
Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Western Blot.
Specifications
| Cytosolic beta-Glucosidase/GBA3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9H227 | |
| GBA3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cytosolic beta-Glucosidase/GBA3 Thr13-Leu469 Accession # Q9H227 | |
| 25 μg | |
| Primary | |
| Detects human Cytosolic beta-Glucosidase/GBA3 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 728714 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CBG, CBGL1, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic betaGlucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, GBA3, GLUC, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57733 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction