missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11542-SP
This item is not returnable.
View return policy
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Galectin-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/mL, CyTOF-ready | |
| P17931 | |
| LGALS3 | |
| E. coli-derived recombinant human Galectin-3 Ala2-Ile250 Accession # P17931 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Galectin-3 is observed and no cross-reactivity with recombinant human (rh) Galectin-8 or rhGalectin-9 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 194801 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3958 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction