missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GAB3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7127-SP
This item is not returnable.
View return policy
Description
GAB3 Monoclonal specifically detects GAB3 in Human samples. It is validated for Western Blot.
Specifications
| GAB3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q8WWW8 | |
| GAB3 | |
| E. coli-derived recombinant human GAB3 Leu121-Asn263 Accession # Q8WWW8 | |
| 25 μg | |
| Primary | |
| Detects human GAB-3 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 720306 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DOS/Gab family member 3, Gab3 scaffolding protein, GRB2-associated binder 3, GRB2-associated binding protein 3, GRB2-associated-binding protein 3, Growth factor receptor bound protein 2-associated protein 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 139716 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction