missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DcR3/TNFRSF6B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2135-SP
This item is not returnable.
View return policy
Description
DcR3/TNFRSF6B Monoclonal specifically detects DcR3/TNFRSF6B in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| DcR3/TNFRSF6B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8771 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization | |
| 229812 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| O95407 | |
| TNFRSF6B | |
| Mouse myeloma cell line NS0-derived recombinant human DcR3/TNFRSF6B Val24-His300 Accession # O95407 | |
| 25 μg | |
| Primary | |
| Detects human DcR3/TNFRSF6B in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Fas, rhOPG, rhTNF R2, rhTRAIL R1, rhTRAIL R2, rhTRAIL R3, or rhTRAIL R4 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction