missing translation for 'onlineSavingsMsg'
Learn More

Human Cyclin A1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18786209
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18786209 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18786209 Supplier R&D Systems Supplier No. MAB7046SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cyclin A1 Monoclonal specifically detects Cyclin A1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 722407
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P78396
Gene Alias CCNA1, cyclin A1, cyclin-A1
Gene Symbols CCNA1
Host Species Mouse
Immunogen E. coli-derived recombinant human Cyclin A1 Lys51-Arg220 Accession # P78396
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8900
Test Specificity Detects human Cyclin A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Cyclin A2, B1, B2, C, D1, D2, D3, E1, E2, or recombinant mouse Cyclin A1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.