missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL4/PF4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB795-SP
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| CXCL4/PF4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5196 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 170106 | |
| Western Blot 2 ug/mL, Simple Western 10 ug/mL | |
| P02776.2 | |
| PF4 | |
| E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2 | |
| 25 μg | |
| Primary | |
| Detects human CXCL4/PF4 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse CXCL4/PF4, recombinant human (rh) GRO alpha , or rhGRO beta . | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction