missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human cIAP-2/HIAP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Brand: R&D Systems AF8171-SP
This item is not returnable.
View return policy
Description
cIAP-2/HIAP-1 Polyclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| cIAP-2/HIAP-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AIP1, API2, BIRC3, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, HIAP-1, MALT2, MIHC, RNF49 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 330 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 1-15 μg/mL | |
| U45878 | |
| BIRC3 | |
| E. coli-derived recombinant human cIAP-2/HIAP-1 Asn2-Ser604 Accession # U45878 | |
| 25 μg | |
| Primary | |
| Detects human cIAP-2/HIAP-1 in Western blots. In Western blots, less than 1% cross-reactivity with cIAP-1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction