Learn More
Human CCL8/MCP-2 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF-281-SP
Product Code. 18726197
  • £168.00 / 25µg
Description
CCL8/MCP-2 Polyclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL8/MCP-2
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Goat
Affinity Purified
RUO
6355
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Neutralization 1.5-5.0 ug/mL
P80075
CCL8
E. coli-derived recombinant human CCL8/MCP-2 Gln24-Pro99 Accession # P80075
25 μg
Primary
Detects human CCL8/MCP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing conditions), less than 40% cross-reactivity with recombinant human (rh) HCC-1 is observed, less than 10% cross-reactivity with rhEotaxin is observed, and less than 5% cross-reactivity with rhMCP-1 is observed. Neutralizes the biological activity of rhCCL8. This antibody will not neutralize the biological activity of rhMCP-1, recombinant mouse JE, or rhMCP-3.