missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL15/MIP-1 delta Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF628-SP
This item is not returnable.
View return policy
Description
CCL15/MIP-1 delta Polyclonal specifically detects CCL15/MIP-1 delta in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL15/MIP-1 delta | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 15, Chemokine CC-2, HCC-2, HCC-2Lkn-1, leukotactin 1, Leukotactin-1, LKN1, LKN-1, Macrophage inflammatory protein 5, MIP1 delta, MIP-1 delta, MIP5, MIP-5, MIP-5MIP-1d, Mrp-2b, NCC-3, NCC-3MIP-1D, NCC3MRP-2B, new CC chemokine 3, SCYA15member 15, Small-inducible cytokine A15 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6359 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.5-2.5 ug/mL | |
| Q16663 | |
| CCL15 | |
| E. coli-derived recombinant human CCL15/MIP-1 delta Ser46-Ile113 Accession # Q16663 | |
| 25 μg | |
| Primary | |
| Detects human CCL15/MIP-1 delta in direct ELISAs and Western blots. In direct ELISAs and Western blots, a preference for the 68 aa residue isoform over the 92 aa residue isoform of human CCL15/LKN-1 is shown. Additionally, in direct ELISAs, less than 1% cross-reactivity with with rh6Ckine, rhBLC/BCA1, rhENA78, rhEotaxin, rhEotaxin2, rhFractalkine, rhGCP2, rhGRO alpha , rhGRO beta , rhGRO gamma , rhHCC1, rhHCC4, rhI309,rhIL8, rhMCP1, rhMCP2, rhMCP3, rhMCP4, rhMDC, rhMIG, rhMIP1 alpha , rhMIP1 beta , rhMIP3 alpha , rhMIP3 beta , rhMPIF1, rhNAP2, rhPARC, rhRANTES, rhSDF1 alpha ,rhSDF1 beta ,rhTarc, rhTECK, and rhVIC is observed. Neutralizes the biological activity of the 68 aa isoform of recombinant human (rh) CCL15/LKN-1. It will also neutralize the biological activity of the 92 aa residue isoform of rhCCL15/LKN-1 using a 2-fold higher Ig concentration. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction