missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspase-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB704-SP
This item is not returnable.
View return policy
Description
Caspase-8 Monoclonal specifically detects Caspase-8 in Human samples. It is validated for Western Blot.
Specifications
| Caspase-8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AIS, androgen receptor, CASP8, Caspase8, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, Mch5, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 841 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 84131 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CASP8 | |
| E. coli-derived recombinant human Caspase-8 aa 234-391 | |
| 25 μg | |
| Primary | |
| Detects human Caspase-8 precursor in Western blots and a 42ákDa doublet generated during apoptosis. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction