missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CAMP/LL37/FALL39 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7497-SP
This item is not returnable.
View return policy
Description
CAMP/LL37/FALL39/Cathelicidin Polyclonal specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot.
Specifications
| CAMP/LL37/FALL39/Cathelicidin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 18 kDa cationic antimicrobial protein, CAMP, CAP18, CAP18CRAMP, cathelicidin antimicrobial peptide, CRAMP, FALL39, FALL-39, FALL39CAP-18, hCAP-18, LL37 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 820 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P49913 | |
| CAMP | |
| E. coli-derived recombinant human CAMP/LL37/FALL39 Leu134-Ser170 Accession # P49913 | |
| 25 μg | |
| Primary | |
| Detects human CAMP/LL37/FALL39 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction