missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-I Tubulin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB8527-SP
This item is not returnable.
View return policy
Description
beta I Tubulin Monoclonal specifically detects beta I Tubulin in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| beta I Tubulin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta tubulin 1, class VI, betaI Tubulin, dJ543J19.4, TUBB1, tubulin beta-1 chain, tubulin, beta 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 81027 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 923425 | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9H4B7 | |
| TUBB1 | |
| E. coli-derived recombinant human beta-I Tubulin Met1-His451 Accession # Q9H4B7 | |
| 25 μg | |
| Primary | |
| Detects human beta-I Tubulin in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction