missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS4 MAb (Clone 416610), R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4307-SP
This item is not returnable.
View return policy
Description
ADAMTS4 Monoclonal specifically detects ADAMTS4 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry.
Specifications
| ADAMTS4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS2, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1, ADMP-1EC 3.4.24.82, Aggrecanase 1, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9507 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry | |
| 416610 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O75173 | |
| ADAMTS4 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAMTS4 Phe213-Cys685 Accession # O75173 | |
| 25 μg | |
| Primary | |
| Detects human ADAMTS4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ADAMTS1, rhADAMTS5, rhADAMTS13, or rhADAMTSL1.2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction