missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Phospho-SHP-2 (Y542) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF3790-SP
This item is not returnable.
View return policy
Description
SHP-2/PTPN11 Polyclonal specifically detects SHP-2/PTPN11 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| SHP-2/PTPN11 (p Tyr542) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PTPN11 | |
| Phosphopeptide containing human SHP-2 Y542 site | |
| 25 μg | |
| Primary | |
| Detects human and mouse SHP-2 when phosphorylated at Y542. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunocytochemistry 3-15 μg/mL | |
| BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP1D, PTP-1D, PTP2C, PTP-2C, PTP2CNS1, PTPN11, SHP2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 5781 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction