missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Phospho-PRAS40 (T246) Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB6890-SP
This item is not returnable.
View return policy
Description
PRAS40 Monoclonal specifically detects PRAS40 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| PRAS40 (p thr246) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| 40 kDa, AKT1 substrate 1 (proline-rich), AKT1S1, Lobe, MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84335 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 760502 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AKT1S1 | |
| Phosphopeptide containing the human PRAS40 T246 site | |
| 25 μg | |
| Primary | |
| Detects human and mouse PRAS40 when phosphorylated at T246 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction